J.T.Baker®, Multipurpose Alkaline Detergent, RBS 35 concentrate
Catalog # 7348.5000
Supplier: Avantor
Specifications
- Beskrivning:Multipurpose alkaline detergent, RBS 35 concentrate
- Förpackningstyp:PE bottle
- Förp.:5 L
- pH-område:11,8 (2%)
Specifications
About this item
RBS 35 concentrate is used for manual cleaning, soaking and ultrasonic baths. These surfactants are alkaline, foaming and insensitive to water hardness.
- High wetting and emulsifying power
- No risk of calcareous deposits when diluted in tap water
- Total rinse
Suitable for cleaning glassware, laboratory equipments and for cleaning parts made of glass, stainless steel, injection moulds and production equipment.
Typical concentration for use: 2%
Not suitable for glass washer machines.