Dehydrated culture media, R-2A agar suitable for microbiology, NutriSelect® Plus
Katalog # SIAL17209-500G
Leverantör: Merck
CAS-nummer:
Specifikationer
- Förp.:500 g
- Beskrivning:R-2A agar
- Vatten:+
Specifikationer
Om denna produkt
R-2A agar is used for heterotrophic plate count of water samples using longer incubation periods.