Spheres for water baths, Clifton®
Catalog # NICKBP0368
Supplier: NICKEL ELECTRO
Specifications
- Ø:20 mm
- Beskrivning:Spheres for water baths, Clifton®
- Förp.:200
- Vikt:250 g
Specifications
About this item
Hollow spheres are ideal for use as a blanket on the top of open tanks of water. A single layer of spheres covers 91% of the liquid surface trapping air in the gaps which adds to the thermal insulation properties. Spheres have 20 mm Ø.
- Reduces thermal losses
- Reduces steaming and splashing