Digital medical thermometer
Catalog # 620-2814
Supplier: Alla France
Specifications
- Mätområde:+32...42 °C
- Beskrivning:Digital medical thermometer
- Noggrannhet:±0,1 °C
- Förp.:1
Specifications
About this item
Digital medical thermometer with flexible probe and sound signal.