ContactUsLinkComponent
Digital medical thermometer
Digital medical thermometer
Catalog # 620-2814
Supplier:  Alla France
undefined
Digital medical thermometer
Catalog # 620-2814
Supplier:  Alla France
Specifications
  • Mätområde:
    +32...42 °C
  • Beskrivning:
    Digital medical thermometer
  • Noggrannhet:
    ±0,1 °C
  • Förp.:
    1
Specifications
About this item
Digital medical thermometer with flexible probe and sound signal.