Ion exchange resins, Diaion® PA312 (Cl)
Katalog # 46426.A1
Leverantör: Thermo Fisher Scientific
CAS-nummer:
Specifikationer
- Partikelstorlek:300 − 1180 µm
- Förp.:1 kg
- Beskrivning:Ion exchange resin, 6% cross-linked, strongly basic porous type, 1,2 meq/ml on PS-DVB
Specifikationer
Om denna produkt
These resins are ideal for use in decolourisation in sugar solutions, metal recovery, pharmaceuticals and purification of amino acids.