Swabs in tubes, Dryswab™
Katalog # MEDWMW1028D
Leverantör: MEDICAL WIRE EQUIPMENT
Specifikationer
- Steril:+
- Beskrivning:Swabs in tubes, Dryswab™
- Längd:150 mm
- Material:Polyester
- Seq:02
- Lock, färg:Svart
- Förp.:800
Specifikationer
Om denna produkt
Individual labelled sterile tubes with polyester bud, standard plastic shaft.
- Tamper-evident label is clearly printed
- Lot number and expiry date included
Recommended for clinical use, forensics, as well as environmental tests.
Shelf life of five years.
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904