Knee pad, KP40
Katalog # PRTWKP40BKR
Leverantör: Portwest
Specifikationer
- Färg:Svart/blå
- Beskrivning:KP40
- Förp.:1 PAIR
Specifikationer
Om denna produkt
This strap-on gel knee pad offers tough protection for gruelling deep knee work. The thick gel adapts to the wearers knee, providing unrivalled comfort and protection.
- Adjustable neoprene strap allows for greater flexibility
- Abrasion resistant PVC shell ensures a longer useful lifespan
- Specially designed shell gives great stability by distributing the weight evenly over a wide area
Certifications: EN 14404:2004+A1:2010 Type 1 Level 1.