Order Entry
Sweden
ContactUsLinkComponent
Anti-VWF Polyclonal Antibody
Anti-VWF Polyclonal Antibody
Katalog # BSBTPB9273
Leverantör:  Boster Bio
undefined
Anti-VWF Polyclonal Antibody
Katalog # BSBTPB9273
Leverantör:  Boster Bio

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Von Willebrand Factor
  • Antigensymbol:
    VWF
  • Klonalitet:
    Polyclonal
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

Polyclonal antibody for VON WILLEBRAND FACTOR/VWF detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Mouse. VON WILLEBRAND FACTOR/VWF information: Molecular Weight: 309269 MW; Subcellular Localization: Secreted . Secreted, extracellular space, extracellular matrix . Localized to storage granules; Tissue Specificity: Plasma. Expressed in liver.

Type: Primary
Antigen: VWF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat