Anti-VE Cadherin Polyclonal Antibody
Catalog # BSBTA02632-2
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:VE Cadherin
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Rat,Mouse
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for VE Cadherin detection. Tested with WB, IHC-P, ELISA(Cap) in Mouse;Rat.
Type: Primary
Antigen: VE Cadherin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat