Anti-Rab3A Polyclonal Antibody
Catalog # BSBTPB9322
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antigensymbol:Rab3A
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Polyclonal antibody for RAB3A detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. RAB3A information: Molecular Weight: 24984 MW; Subcellular Localization: Cell membrane ; Lipid-anchor ; Cytoplasmic side ; Tissue Specificity: Specifically expressed in brain.
Type: Primary
Antigen: Rab3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat