- Antikroppstyp:Primär
- Antikroppsnamn:PKC Epsilon
- Antigensymbol:PKCE
- Klonalitet:Polyclonal
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Polyclonal antibody for PKC epsilon/PRKCE detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. PKC epsilon/PRKCE information: Molecular Weight: 83674 MW; Subcellular Localization: Cytoplasm . Cytoplasm, cytoskeleton . Cell membrane . Cytoplasm, perinuclear region . Nucleus . Translocated to plasma membrane in epithelial cells stimulated by HGF. Associated with the Golgi at the perinuclear site in pre-passage fibroblasts (By similarity). In passaging cells, translocated to the cell periphery (By similarity). Translocated to the nucleus in PMA- treated cells (By similarity).
Type: Primary
Antigen: PKC Epsilon
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat