Order Entry
Sweden
ContactUsLinkComponent
Anti-Neuregulin-1 Polyclonal Antibody
Anti-Neuregulin-1 Polyclonal Antibody
Catalog # BSBTPB9299
Supplier:  Boster Bio
undefined
Anti-Neuregulin-1 Polyclonal Antibody
Catalog # BSBTPB9299
Supplier:  Boster Bio

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Neuregulin-1
  • Klonalitet:
    Polyclonal
  • Värd:
    Rabbit
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifications

About this item

Polyclonal antibody for Neuregulin 1/NRG1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Neuregulin 1/NRG1 information: Molecular Weight: 70392 MW; Subcellular Localization: Pro-neuregulin-1, membrane-bound isoform: Cell membrane; Single-pass type I membrane protein. Does not seem to be active; Tissue Specificity: Type I isoforms are the predominant forms expressed in the endocardium. Isoform alpha is expressed in breast, ovary, testis, prostate, heart, skeletal muscle, lung, placenta liver, kidney, salivary gland, small intestine and brain, but not in uterus, stomach, pancreas, and spleen. Isoform 3 is the predominant form in mesenchymal cells and in non-neuronal organs, whereas isoform 6 is the major neuronal form. Isoform 8 is expressed in spinal cord and brain. Isoform 9 is the major form in skeletal muscle cells; in the nervous system it is expressed in spinal cord and brain. Also detected in adult heart, placenta, lung, liver, kidney, and pancreas. Isoform 10 is expressed in nervous system: spinal cord motor neurons, dorsal root ganglion neurons, and brain. Predominant isoform expressed in sensory and motor neurons. Not detected in adult heart, placenta, lung, liver, skeletal muscle, kidney, and pancreas. Not expressed in fetal lung, liver and kidney. Type IV isoforms are brain-specific.

Type: Primary
Antigen: Neuregulin-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human