Anti-IP3 Receptor Polyclonal Antibody
Katalog # BSBTPB9225
Leverantör: Boster Bio
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:IP3 Receptor
- Klonalitet:Polyclonal
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Polyclonal antibody for IP3R1/ITPR1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. IP3R1/ITPR1 information: Molecular Weight: 313929 MW; Subcellular Localization: Endoplasmic reticulum membrane; Multi-pass membrane protein; Tissue Specificity: Widely expressed.
Type: Primary
Antigen: IP3 Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat