Anti-Integrin Alpha 5 Polyclonal Antibody
Catalog # BSBTPB9254
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Integrin Alpha 5
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Polyclonal antibody for CD49e/ITGA5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. CD49e/ITGA5 information: Molecular Weight: 114536 MW; Subcellular Localization: Membrane; Single-pass type I membrane protein.
Type: Primary
Antigen: Integrin Alpha 5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat