Order Entry
Sweden
ContactUsLinkComponent
Anti-IL27 Polyclonal Antibody
Anti-IL27 Polyclonal Antibody
Katalog # BSBTA00857-2
Leverantör:  Boster Bio
undefined
Anti-IL27 Polyclonal Antibody
Katalog # BSBTA00857-2
Leverantör:  Boster Bio

Specifikationer

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    IL27
  • Klonalitet:
    Polyclonal
  • Värd:
    Rabbit
  • Reaktivitet:
    Rat,
    Mouse
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

Rabbit IgG polyclonal antibody for IL27 detection. Tested with WB, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: IL27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse; Rat