Anti-hnRNP A2B1 Polyclonal Antibody
Catalog # BSBTA00396-1
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antigensymbol:hnRNP A2B1
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for hnRNP A2B1 detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: hnRNP A2B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat