Anti-FABP5 Polyclonal Antibody
Katalog # BSBTA02083
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antigensymbol:FABP5
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for FABP5 detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: FABP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat