Anti-c-Myb Polyclonal Antibody
Catalog # BSBTPB9288
Leverantör: Boster Bio
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:c-Myb
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Polyclonal antibody for C MYB/MYB detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. C MYB/MYB information: Molecular Weight: 72341 MW; Subcellular Localization: Nucleus .
Type: Primary
Antigen: c-Myb
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human