Order Entry
Sweden
ContactUsLinkComponent
Anti-ATF1 Polyclonal Antibody
Anti-ATF1 Polyclonal Antibody
Catalog # BSBTPB9130
Supplier:  Boster Bio
undefined
Anti-ATF1 Polyclonal Antibody
Catalog # BSBTPB9130
Supplier:  Boster Bio

Specifications

  • Antikroppstyp:
    Primär
  • Antigensymbol:
    ATF1
  • Klonalitet:
    Polyclonal
  • Flödescytometri:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Human,
    Rat
  • Western blot:
    Yes
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifications

About this item

Polyclonal antibody for 19261910 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. 19261910 information: Molecular Weight: 29233 MW; Subcellular Localization: Nucleus.

Type: Primary
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat