Weighing dishes, aluminium
Katalog # MARI6604900
Leverantör: MARIENFELD
Specifikationer
- Ø utv.:100 mm
- Beskrivning:Weighing dishes, 80 per pack, 6 packs
- Höjd:7 mm
- Förp.:80
Specifikationer
Om denna produkt
Dishes made of aluminium.
- Flat bottom ensures good standing on the worktop and keeps the samples securely in place
- Manufactured free of grease or oil
- Good chemical and temperature resistance
Suitable for moisture analysis in infra-red dryer.