Human TTYH2 peptide
Katalog # PRSI6885P
Leverantör: ProSci Inc.
CAS-nummer:
Specifikationer
- Förp.:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Aminosyretal:14 amino acid peptide near the amino terminus of human TTYH2
- Protein/peptide name:TTYH2 Peptide
- Tested applications:Blocking
Specifikationer
Om denna produkt
TTYH2 peptide is used for blocking the activity of TTYH2 antibody.