Specifications
- Antikroppsnamn:Green Fluorescence Protein
- Antigensymbol:GFP
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Western blot:Yes
- Form:Liquid
- Antigen synonymer:GFPuv|eGFP|Green Fluorescent Protein
- Koncentration:1.0 mg/mL
- Immunogen:Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Polyclonal anti-GFP is designed to detect GFP and its variants. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen and recognizes wild type, recombinant and enhanced forms of GFP. For immunoblotting use either alkaline phosphatase or peroxidase conjugated polyclonal anti-GFP to detect GFP or GFP containing proteins on western blots. Recommended dilutions are as follows, ELISA 1:15,000 - 1:60,000, Western Blot 1:500 - 1:5,000, Immunohistochemistry 1:200 - 1:3,000. (Optimal dilutions should be determined by user).
Anti-GFP Antibody was prepared from monospecific antiserum by immunoaffinity chromatography using Green Fluorescent Protein (Aequorea victoria) coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum and purified and partially purified Green Fluorescent Protein (Aequorea victoria). No reaction was observed against Human, Mouse or Rat serum proteins.
0.02 M Potassium Phosphate, 0.15 M NaCl, 0.01% (w/v) Sodium Azide; pH 7.2
Store vial at -20° C or below prior to opening. Dilute only prior to immediate use. Forextended storage, aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Expiration date is one (1) year from date of opening product.
Type:
Antigen: gfp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: GFP