Specifikationer
- Antikroppsnamn:Glutamate Ionotropic Receptor NMDA Type Subunit 2B
- Antigensymbol:GRIN2B
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonymer:hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B|glutamate ionotropic receptor NMDA type subunit 2B
- Immunogen:NMDA NR2B (Tyr1472) polyclonal antibody was raised against a synthetic phosphopeptide corresponding to amino acids residues surrounding the phosphorylated Tyr1472 of rat NMDA NR2B.
- Storlek:100 µl
- Förp.:100 µl
Specifikationer
Om denna produkt
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR). The NMDAR plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimer’s, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity.
The labeling of the NR2B protein in Western blots of rat brain is blocked by the phosphopeptide but not by the dephosphopeptide. The antibody also labels bands at 65k and 115k. Applications include Dot Blots (DB) and Western Blots (WB). Human, mouse, and rat have 100% amino acid sequence identity with the antigen used to raise the antibody. When internally tested under ideal conditions the working dilutions were 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
For long term storage –80°C is recommended, but shorter term storage at –20°C is also acceptable as aliquots may be taken without freeze/thawing due to the presence of 50% glycerol. Stable for one year.
Type:
Antigen: Grin2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat