Specifikationer
- Antikroppsnamn:Interleukin 6
- Antigensymbol:IL6
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Reaktivitet:Mouse
- Western blot:Yes
- Neutralisation:Yes
- Form:Lyophilized
- Antigen synonymer:HSF|HGF|IFNB2|interferon beta 2|BSF2|IL-6
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mIL-6 (murine Interleukin-6).
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. It acts upon a variety of cells including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. In addition, IL-6 appears to interact with IL2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 is a multifunctional 24 kDa protein originally discovered in the medium of RNA stimulated fibroblastoid cells. It is upregulated by IL1, TNF, PDGF, IFN beta, TNF alpha, NGF, IL17 and downregulated by glucocorticoids IL4, TGF beta. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be as important as IL1 and TNF alpha in regulating the acute phase response. IL6 has also been implicated in regulating adipose mass.
Anti-mIL-6 specific antibody was purified by affinity chromatography employing immobilized mIL-6 matrix.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse