- Antikroppsnamn:TBC1 Domain Family Member 4
- Antigensymbol:TBC1D4
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human
- Form:Liquid
- Antigen synonymer:CDC16) domain-containing protein|Acrg embryonic lethality minimal region ortholog|AS160|TBC1 domain family member 4|akt substrate of 160 kDaStore at 4║C for 1-2 weeks. For long-term storage|store at -20║C.|TBC (Tre-2|RP11-159J2.3|DKFZp779C0666|BUB2
- Koncentration:1 mg/mL
- Immunogen:TBC1D4 antibody was raised against an 19 amino acid peptide near the carboxy terminus of human TBC1D4 .
- Storlek:100 µg
- Förp.:100 µG
TBC1D4 Antibody: TBC1D4, also known as the Akt substrate AS160, was initially identified as an Akt substrate containing a GTPase-activating domain that regulates GLUT4 trafficking, with activation following insulin stimulation. TBC1D4 truncations in humans is a major cause of dominant inherited insulin resistance. The loss of TBC1D4 results in the accumulation of GLUT4 in compartments that are primed for fusion in basal adipocytes.
TBC1D4 Antibody is affinity chromatography purified via peptide column.
TBC1D4 Antibody is supplied in PBS containing 0.02% sodium azide.
TBC1D4 antibody can be stored at 4°C for three months and -20°C, stable for up to one year.
Type:
Antigen: TBC1D4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human