Anti-PRDM14 Goat Polyclonal Antibody
Catalog # PRSI42-423
Supplier: ProSci Inc.
Specifications
- Antikroppsnamn:PR Domain Zinc Finger Protein 14
- Antigensymbol:PRDM14
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Goat
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonymer:PR domain-containing protein 14|PFM11|PR domain zinc finger protein 14
- Koncentration:500 ug/mL
- Immunogen:PRDM14 antibody was raised against a 14 amino acid synthetic peptide near the internal region of PRDM14 (aa 414-427).
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
PRDM14 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
PRDM14 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PRDM14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat