Anti-Respiratory Syncytial Virus Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI35-350
Supplier: ProSci Inc.
Specifications
- Antikroppsnamn:Respiratory Syncytial Virus
- Antigensymbol:RSV
- Klonalitet:Polyclonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Värd:Goat
- ImmunKemi:Yes
- Reaktivitet:Virus
- Western blot:Yes
- Form:Liquid
- Antigen synonymer:RSV|human respiratory syncytial virus
- Immunogen:Human RSV isolate, confirmed
- Storlek:1 mL
- Förp.:1 mL
Specifications
About this item
Purified IgG fraction of antiserum covalently coupled with high purity Isomer I of fluorescein isothiocyanate. Care is taken to insure complete removal of any free fluorescein from the final product.
Respiratory Syncytial Virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing BSA (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Respiratory Syncytial Virus
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Goat
Isotype:
Reactivity: Virus