Anti-ZFP42 Rabbit Polyclonal Antibody
Catalog # PRSI28-468
Supplier: ProSci Inc.
Specifications
- Antikroppsnamn:Zinc Finger Protein 42
- Antigensymbol:ZFP42
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonymer:zfp-42|REX1|ZNF754|REX-1
- Immunogen:A synthetic peptide corresponding a region of human ZFP42.
- Storlek:50 µg
- Förp.:50 µG
Specifications
About this item
ZFP42 is a transcription factors and a specific marker of undifferentiated embryonic stem (ES) cells.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZFP42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human