Anti-SMYD3 Rabbit Polyclonal Antibody
Katalog # PRSI26-556
Leverantör: ProSci Inc.
Specifikationer
- Antikroppsnamn:SET And MYND Domain Containing 3
- Antigensymbol:SMYD3
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonymer:Zinc finger MYND domain-containing protein 1|SMYD3|ZMYND1|ZNFN3A1
- Immunogen:A synthetic peptide corresponding a region of human SMYD3.
- Storlek:50 µg
- Förp.:50 µG
Specifikationer
Om denna produkt
SMYD3 is a member of an RNA polymerase complex. Within the RNA polymerase complex, it acts as a histone methyltransferase that plays a role in transcriptional regulation.SMYD3 is a histone methyltransferase that plays a role in transcriptional regulation as a member of an RNA polymerase complex.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SMYD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat