Specifikationer
- Antikroppsnamn:Oculocutaneous albinism 2
- Antigensymbol:OCA2
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- Reaktivitet:Human,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonymer:EYCL2|EYCL|P|EYCL3|BEY|BEY2|D15S12|BOCA|BEY1|PED|SHEP1|HCL3
- Immunogen:A synthetic peptide corresponding a region of human OCA2.
- Storlek:50 µg
- Förp.:50 µG
Specifikationer
Om denna produkt
OCA2 is believed to be an integral membrane protein involved in small molecule transport, specifically tyrosine - a precursor of melanin. Mutations in the gene encoding OCA2 result in type 2 oculocutaneous albinism.This gene encodes the human homologue of the mouse p (pink-eyed dilution) gene. The encoded protein is believed to be an integral membrane protein involved in small molecule transport, specifically tyrosine - a precursor of melanin. Mutations in this gene result in type 2 oculocutaneous albinism. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: OCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse