- Antikroppsnamn:Tumor Endothelial Marker 5
- Antigensymbol:TEM5
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat
- Western blot:Yes
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:TEM5 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human TEM5.
- Tested applications:E, WB, IHC
- Rening:Affinity chromatography purified via peptide column
- Storlek:100 µg
- Förp.:100 µG
TEM5 Antibody: Tumor endothelial markers (TEMs) are significantly up-regulated during angiogenesis and neoangiogensis that are crucial for the growth of solid tumors. TEMs localized on the cell surface and conserved across species are of particular interest for future development of anti-angiogenic therapies. These include TEMs such as TEM1, TEM5, TEM7 and TEM8. TEM5 is a member of the adhesion family of G protein coupled receptors and is localized on the surface of endothelial cells. TEM5 is a seven-pass transmembrane receptor, unlike TEM1, TEM7 and TEM8 which span the membrane once. TEM5 is abundantly expressed in tumor vessels, heart, placenta, ovary, small intestine, and colon. Proteolytically processed soluble TEM5 mediates endothelial cell survival during angiogenesis by linking integrin to glycosaminoglycans.
Type:
Antigen: TEM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat