Specifikationer
- Antikroppsnamn:Caspase 1
- Antigensymbol:CASP1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Antigen synonymer:interleukin 1-B converting enzyme|ICE|IL1B-convertase|P45|apoptosis-related cysteine peptidase interleukin 1|IL1BC|beta|IL-1 beta-converting enzyme|caspase 1|interleukin 1|convertase|caspase-1|CASP1 nirs variant 1
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:Caspase-1 antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human Caspase-1.
- Tested applications:E, WB, IHC, IF
- Rening:Affinity chromatography purified via peptide column
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Caspase-1 Antibody: Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1 beta and IL-18. Caspase-1 was initially identified as an IL-1 beta -converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.
Type:
Antigen: CASP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human