Elemental analysis standards, portland cement (Ivory)
Catalog # PROONIST-1884B
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Förp.:5
- Beskrivning:Portland cement (Ivory) NIST-1884B
- Pk Info:5 x 4,5 g
Specifications
About this item
These are intended primarily for the evaluation and calibration of methods for analysis of cements and materials of similar matrix.
A unit consists of five sealed vials, each containing approximately 4,5 g of portland cement ground to pass through a 75 µm (No. 200) sieve.
Certifications: Traceable to NIST and SRM.