Anti-Ace Rabbit Polyclonal Antibody
Catalog # BSBTA00251
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antigensymbol:Ace
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Rat,Mouse
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Ace detection. Tested with WB in Mouse;Rat.
Type: Primary
Antigen: Ace
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat