Anti-VEGFB Rabbit Polyclonal Antibody
Katalog # BSBTA04494-1
Leverantör: Boster Bio
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:VEGFB
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Rabbit IgG polyclonal antibody for VEGFB detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: VEGFB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat