Anti-Bid Rabbit Polyclonal Antibody
Katalog # BSBTA00730
Leverantör: Boster Bio
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:BH3-interacting domain death agonist
- Antigensymbol:Bid
- Klonalitet:Polyclonal
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Rabbit IgG polyclonal antibody for BH3-interacting domain death agonist(Bid) detection. Tested with WB, IHC-P in Mouse;Rat.
Type: Primary
Antigen: Bid
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat