Anti-PDGFB Rabbit Polyclonal Antibody
Catalog # BSBTA00348
Leverantör: Boster Bio
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Platelet-derived growth factor subunit B
- Antigensymbol:PDGFB
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Rabbit IgG polyclonal antibody for Platelet-derived growth factor subunit B(PDGFB) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: PDGFB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat