Anti-FDCSP Rabbit Polyclonal Antibody
Catalog # BSBTA13093-1
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Follicular dendritic cell secreted peptide
- Antigensymbol:FDCSP
- Klonalitet:Polyclonal
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Human
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Follicular dendritic cell secreted peptide(FDCSP) detection. Tested with IHC-P in Human.
Type: Primary
Antigen: FDCSP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human