Anti-EIF3E Rabbit Polyclonal Antibody
Catalog # BSBTA00481-1
Supplier: Boster Bio
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:Eukaryotic translation initiation factor 3 subunit E
- Antigensymbol:EIF3E
- Klonalitet:Polyclonal
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Eukaryotic translation initiation factor 3 subunit E(EIF3E) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: EIF3E
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat