Anti-CHAT Rabbit Polyclonal Antibody
Katalog # BSBTA01192-3
Leverantör: Boster Bio
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Choline O-acetyltransferase
- Antigensymbol:CHAT
- Klonalitet:Polyclonal
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Rabbit IgG polyclonal antibody for Choline O-acetyltransferase(CHAT) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: CHAT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat