Orders LinkContactUsLinkComponent
Propofol, Reference Standards
Propofol, Reference Standards
Leverantör:   USP
Propofol, Reference Standards
Leverantör:  USP
Certificates
2 Alternativ tillgängliga nedan
Produktdetaljer och dokument
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Dokument
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • 1572525.
  • 1572503.
  • Benämning:
  • Propofol Resolution Mixture
  • Propofol
  • Synonym(s):
  • Propofol, 2,6-diisopropylphenyl isopropylether, and 2-isopropyl-6-n-propylphenol
  • CAS-nr:
  • 2078-54-8
Specifikationer
Product Family Options
Produktinformation
  • Standard
    Förpackningstyp
    Förp.
    Tillgänglighet
    Price
  • 1572525.
    Propofol Resolution Mixture
    Propofol Resolution Mixture
    USP reference standard
    Ampule
    100 mg
    Log in för att kolla tillgänglighet och ert pris
    Log in för att kolla tillgänglighet och ert pris
    1 * 100 mg
    1 * 100 mg - 14 550,00 kr
    14 550,00 kr
    1 * 100 mg
       14 550,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1572525.
    Benämning
    Propofol Resolution Mixture
    Synonym(s)
    Propofol, 2,6-diisopropylphenyl isopropylether, and 2-isopropyl-6-n-propylphenol
    CAS-nr
  • Propofol
    USP reference standard
    Ampule
    500 mg
    Log in för att kolla tillgänglighet och ert pris
    Log in för att kolla tillgänglighet och ert pris
    1 * 500 mg
    1 * 500 mg - 4 575,00 kr
    4 575,00 kr
    1 * 500 mg
       4 575,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1572503.
    Benämning
    Propofol
    Synonym(s)
    CAS-nr
    2078-54-8