Quartz wool
Quartz wool, loosely packed 2 - 12 µm fibres, 500 g
Katalog # QUTW1186
Leverantör: QUARTZTECHNISCHE WERKSTATTEN
Gå till:
Product Details & Documents
Loosely, fibre size 2 to12 μm.
Documents
TDS Quartz Wool.pdf
Datablad
Specifikationer
- Beskrivning:Quartz wool, loosely packed 2 - 12 µm fibres, 500 g
- Seq:01