Anti-THBS1 Rabbit Polyclonal Antibody
BSBTA00667-1
- Antikroppstyp:Primär
- Antikroppsnamn:Thrombospondin-1
- Antigensymbol:THBS1
- Klonalitet:Polyclonal
- ELISA:Yes
- Värd:Rabbit
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Rabbit IgG polyclonal antibody for Thrombospondin-1(THBS1) detection. Tested with WB, ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: THBS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat