Order Entry
Sweden
ContactUsLinkComponent
Anti-PPIA Rabbit Polyclonal Antibody
Anti-PPIA Rabbit Polyclonal Antibody
Katalog # BSBTA01308
Leverantör:  Boster Bio
Anti-PPIA Rabbit Polyclonal Antibody
Katalog # BSBTA01308
Leverantör:  Boster Bio

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Peptidyl-prolyl cis-trans isomerase A
  • Antigensymbol:
    PPIA
  • Klonalitet:
    Polyclonal
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

Rabbit IgG polyclonal antibody for Peptidyl-prolyl cis-trans isomerase A(PPIA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: PPIA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat