Domestic sludge - metals, NIST-2781 standard
Katalog # PROONIST-2781
Leverantör: LGC Standards PROMOCHEM
CAS-nummer:
Specifikationer
- Förp.:40 g
- Beskrivning:NIST-2781, domestic sludge - metals
Specifikationer
Om denna produkt
This standard reference material is intended to use in the evaluation of methods used for the analysis of sludges and other materials of a similar matrix. These are dried and pulverised domestic sludge.
A unit consists of 40 g of the dried material.
Certifications: Traceable to NIST and SRM numbers.