Baking chocolate standard, NIST-2384
Katalog # PROONIST-2384
Leverantör: LGC Standards PROMOCHEM
CAS-nummer:
Specifikationer
- Förp.:5
- Beskrivning:NIST-2384, baking chocolate
Specifikationer
Om denna produkt
This reference material intended to use in validating methods for determining proximates, fatty acids, calories, vitamins, elements, catechins, caffeine, theobromine, and acrylamide in baking chocolate and similar matrices. It can be used for quality assurance in-house control materials.
A unit consists of five 91 g individually wrapped bars of baking chocolate.
Certifications: Traceable to NIST and SRM.