Whole milk powder, NIST-1549A
Katalog # PROONIST-1549A
Leverantör: LGC Standards PROMOCHEM
CAS-nummer:
Specifikationer
- Förp.:5
- Beskrivning:NIST-1549A, whole milk powder
- Pk Info:5× 10 g sealed aluminised pouches
Specifikationer
Om denna produkt
This reference standard is intended for validation of methods for determining proximates, fatty acids, cholestrol, vitamins, elements and amino acids in whole milk powder and similar materials.
Certifications: Traceable to NIST and SRM numbers.