ContactUsLinkComponent
Regenerating salts for dishwashers
Regenerating salts for dishwashers
Catalog # 148-2501
Supplier:  KAMMERER
Regenerating salts for dishwashers
Catalog # 148-2501
Supplier:  KAMMERER
Specifications
  • Beskrivning:
    Regenerating salt, 2 kg
  • Förp.:
    2 kg
Specifications
About this item
Regenerating salts for water softening systems for dishwashers, it protects dishes and machines from calcification. Also they ensures optimum efficiency of the dish cleaner and are meant for all cleaning systems.
  • Highly pure and residue-free soluble
  • Coarse-grained for optimal solubility
Certifications: EN 973 Type A.