VWR®, Emergency Intervention Kit
129-0199
- Beskrivning:Emergency intervention kit
- Förp.:1 SET
Set for absorption of organics, aqueous solutions, alkalis and acids. It contains.
- 3× MERCK Chemizorb for absorption of organics and aqueous solutions (1.02051.0500)
- MERCK Chemizorb for absorption of alkalis (including a neutraliser and a pH indicator) (1.01596.1000)
- Absorbent PIG absorbandlock acid PLP501
- Pair of barrier gloves with liner size 9 (112-1010)
- Pair of barrier gloves with liner size 7 (112-0946)
- 2× masks 4279 3M ABEK (111-0044)
- 2× goggles 3M 2890A (111-0434)
- Trash tray ergonomic (129-0174)
- Brush in PBT, sterilisable (129-0175)
- Disposal bag 483×610 mm (129-0021)
- PP box
This product is not available in all countries. Please check with your local Avantor office or supplier.