Chemical resistant gloves, UltraNeo 339
Leverantör: MAPA
Certifikat
Om denna produkt
These neoprene gloves with textile lining provide superior chemical resistance against acids and alkalis, such as concentrated hydrofluoric acid, oils, and greases.
- 100% neoprene multi-layer construction
- Pebbled embossing for improved grip on wet and slippery objects
- Excellent mechanical resistance and longer service life thanks to the textile lining and the thickness of the glove
- Cotton knit lining with thermal insulaton: Suitable for handling hot liquids
Designed using an exclusive anatomical shape, ChemZoil gloves offer excellent fit and exceptional comfort, even when worn for long periods. The pre-curved fingers provide unrivalled freedom of movement.
Typical applications include: Surface treatment of metals (acids), site pollution control, resin-working, petrochemical product sampling, handling on alkylation units, handling and transporting drums of chemical products.
Certifications: EN 388:2016 (3121X), EN ISO 374-1 Type A (ABCJLMNS), EN 407 (X1XXXX); Cat. III
Specifikationer
- Catalog No:
- 112-0665
- 112-0668
- Modell:
- UltraNeo 339
- UltraNeo 339
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Yta:
- Pebbled
- Pebbled
- Kragmodell:
- Sicksackskuren kant
- Sicksackskuren kant
- EN standard:
- EN 388:2016 (3121X), EN ISO 374-1 Type A (ABCJLMNS), EN 407 (X1XXXX)
- EN 388:2016 (3121X), EN ISO 374-1 Type A (ABCJLMNS), EN 407 (X1XXXX)